Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nsa2
Entrez Gene ID
321769
Gene Name
NSA2 ribosome biogenesis homolog (S. cerevisiae)
Gene Aliases
fb34b10, wu:fb34b10, zgc:56334
Chromosome Location
Chr.21: 3789839 - 3796480 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199568.2 NP_955862.2 5 - 6 904 96
GenBank mRNA BC046083.1 - 5 - 6 853 96
EH457038.1 - 5 - 6 893 96
EH531450.1 - 1 - 2 280 96

View Full Product Details