Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk20
Entrez Gene ID
494112
Gene Name
cyclin-dependent kinase 20
Gene Aliases
zgc:101530
Chromosome Location
Chr.8: 7494941 - 7504201 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008655.1 NP_001008655.1 5 - 6 792 86
XM_005166689.4 XP_005166746.1 6 - 7 672 86
XM_009303748.3 XP_009302023.1 6 - 7 670 86
XM_021478244.1 XP_021333919.1 5 - 6 780 86
GenBank mRNA BC086697.1 - 5 - 6 792 86
BC164595.1 - 3 - 4 350 86

View Full Product Details