Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tab3
Entrez Gene ID
566547
Gene Name
TGF-beta activated kinase 1 (MAP3K7) binding protein 3
Gene Aliases
map3k7ip3, map3k7ip3l, si:ch211-232d19.8, si:dkey-30h22.6
Chromosome Location
Chr.8: 18545490 - 18562979 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045105.1 NP_001038570.1 6 - 7 1807 119
XM_017357352.2 XP_017212841.1 6 - 7 2115 119
GenBank mRNA BC134845.1 - 6 - 7 1718 119

View Full Product Details