Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jak2a
Entrez Gene ID
30307
Gene Name
Janus kinase 2a
Gene Aliases
jak4
Chromosome Location
Chr.21: 249819 - 269898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131093.1 NP_571168.1 6 - 7 736 99
XM_009295057.3 XP_009293332.1 6 - 7 99
GenBank mRNA AJ005690.1 - 6 - 7 736 99
BC162596.1 - 6 - 7 695 99

View Full Product Details