Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nucb2a
Entrez Gene ID
373078
Gene Name
nucleobindin 2a
Gene Aliases
cb638, nucb2, wu:fa03d10
Chromosome Location
Chr.7: 56540 - 69730 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201493.1 NP_958901.1 9 - 10 1012 75
GenBank mRNA BC046077.1 - 9 - 10 1012 75
BC065437.1 - 9 - 10 978 75

View Full Product Details