Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fibpb
Entrez Gene ID
405787
Gene Name
fibroblast growth factor (acidic) intracellular binding protein b
Gene Aliases
fibpl, zgc:103503
Chromosome Location
Chr.21: 25589121 - 25613249 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212861.2 NP_998026.1 10 - 11 1210 69
XM_009295296.3 XP_009293571.1 10 - 11 1331 69
XM_009295297.3 XP_009293572.1 10 - 11 1106 69
GenBank mRNA AY427781.1 - 9 - 10 1006 69
BC076477.1 - 10 - 11 1271 69
BC085535.1 - 10 - 11 1210 69

View Full Product Details