Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fibpb
Entrez Gene ID
405787
Gene Name
fibroblast growth factor (acidic) intracellular binding protein b
Gene Aliases
fibpl, zgc:103503
Chromosome Location
Chr.21: 25589121 - 25613249 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212861.2 NP_998026.1 3 - 4 490 82
XM_009295296.3 XP_009293571.1 3 - 4 611 82
XM_009295297.3 XP_009293572.1 3 - 4 386 82
XM_021468797.1 XP_021324472.1 3 - 4 611 82
XM_021468798.1 XP_021324473.1 3 - 4 611 82
GenBank mRNA AY427781.1 - 2 - 3 286 82
BC076477.1 - 3 - 4 535 82
BC085535.1 - 3 - 4 490 82

View Full Product Details