Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fibpb
Entrez Gene ID
405787
Gene Name
fibroblast growth factor (acidic) intracellular binding protein b
Gene Aliases
fibpl, zgc:103503
Chromosome Location
Chr.21: 25589121 - 25613249 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212861.2 NP_998026.1 6 - 7 851 144
XM_009295296.3 XP_009293571.1 6 - 7 972 144
XM_009295297.3 XP_009293572.1 6 - 7 747 144
XM_021468797.1 XP_021324472.1 6 - 7 972 144
XM_021468798.1 XP_021324473.1 6 - 7 972 144
GenBank mRNA AY427781.1 - 5 - 6 647 144
BC076477.1 - 6 - 7 912 144
BC085535.1 - 6 - 7 851 144

View Full Product Details