Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fibpb
Entrez Gene ID
405787
Gene Name
fibroblast growth factor (acidic) intracellular binding protein b
Gene Aliases
fibpl, zgc:103503
Chromosome Location
Chr.21: 25589121 - 25613249 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212861.2 NP_998026.1 9 - 10 1110 130
XM_009295296.3 XP_009293571.1 9 - 10 1231 130
XM_009295297.3 XP_009293572.1 9 - 10 1006 130
XM_021468797.1 XP_021324472.1 9 - 10 1231 130
XM_021468798.1 XP_021324473.1 9 - 10 1231 130
GenBank mRNA AY427781.1 - 8 - 9 906 130
BC076477.1 - 9 - 10 1171 130
BC085535.1 - 9 - 10 1110 130

View Full Product Details