Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd10b
Entrez Gene ID
405869
Gene Name
programmed cell death 10b
Gene Aliases
zgc:85629
Chromosome Location
Chr.2: 36721459 - 36731963 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212933.1 NP_998098.1 6 - 7 591 109
GenBank mRNA BC067550.1 - 6 - 7 591 109

View Full Product Details