Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plin2
Entrez Gene ID
794841
Gene Name
perilipin 2
Gene Aliases
adfp, zgc:110411
Chromosome Location
Chr.15: 154546 - 165901 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030262.1 NP_001025433.1 4 - 5 375 65
GenBank mRNA BC096807.1 - 4 - 5 375 65
BC164524.1 - 3 - 4 337 65

View Full Product Details