Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp23
Entrez Gene ID
100004177
Gene Name
ATP23 metallopeptidase and ATP synthase assembly factor homolog
Gene Aliases
xrcc6bp1, zgc:162885
Chromosome Location
Chr.4: 958821 - 965458 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083580.1 NP_001077049.1 5 - 6 744 98
GenBank mRNA BC135073.1 - 5 - 6 744 98

View Full Product Details