Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
laptm4b
Entrez Gene ID
368911
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
fd60g10, si:zc215i13.7, wu:fd60g10, zgc:73120
Chromosome Location
Chr.19: 1227 - 25408 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200098.2 NP_956392.1 5 - 6 695 65
GenBank mRNA BC059484.1 - 5 - 6 624 65
EH440716.1 - 5 - 6 668 65

View Full Product Details