Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl32
Entrez Gene ID
751730
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
zgc:153328
Chromosome Location
Chr.16: 43345162 - 43346956 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045440.2 NP_001038905.2 1 - 2 177 81
GenBank mRNA BC122245.1 - 1 - 2 168 81
EH541038.1 - 1 - 2 216 81
EH565686.1 - 2 - 3 495 81

View Full Product Details