Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aspa
Entrez Gene ID
557232
Gene Name
aspartoacylase
Gene Aliases
ACY-2, zgc:171507
Chromosome Location
Chr.5: 62340799 - 62356967 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110103.1 NP_001103573.1 2 - 3 595 71
GenBank mRNA BC153945.1 - 2 - 3 595 71

View Full Product Details