Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
foxp3a
Entrez Gene ID
571165
Gene Name
forkhead box P3a
Gene Aliases
foxp3
Chromosome Location
Chr.8: 25716307 - 25731177 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001329567.1 NP_001316496.1 7 - 8 708 85
XM_021478298.1 XP_021333973.1 7 - 8 764 85
GenBank mRNA BC154484.1 - 7 - 8 708 85
BC163356.1 - 6 - 7 687 85
FJ906821.1 - 6 - 7 614 85
FM881778.1 - 6 - 7 681 85

View Full Product Details