Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan15
Entrez Gene ID
100000710
Gene Name
tetraspanin 15
Gene Aliases
im:7155083, zgc:110375
Chromosome Location
Chr.13: 22926117 - 22961591 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017773.1 NP_001017773.1 4 - 5 553 108
GenBank mRNA BC092907.1 - 4 - 5 553 108
BC152126.1 - 4 - 5 462 108

View Full Product Details