Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pth2
Entrez Gene ID
402812
Gene Name
parathyroid hormone 2
Gene Aliases
tip39
Chromosome Location
Chr.17: 33495194 - 33499023 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205577.3 NP_991140.1 2 - 3 367 66
GenBank mRNA AF486190.1 - 2 - 3 347 66
AY306196.1 - 2 - 3 228 66
AY307076.1 - 2 - 3 165 66
BC076066.1 - 2 - 3 379 66
BC164665.1 - 1 - 2 138 66

View Full Product Details