Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gtf2h3
Entrez Gene ID
436837
Gene Name
general transcription factor IIH, polypeptide 3
Gene Aliases
fc26a07, gtf2h3-2, wu:fc26a07, zgc:92759
Chromosome Location
Chr.5: 28207 - 38966 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128422.1 NP_001121894.1 5 - 6 529 68
GenBank mRNA BC076236.1 - 5 - 6 501 68

View Full Product Details