Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cant1b
Entrez Gene ID
445201
Gene Name
calcium activated nucleotidase 1b
Gene Aliases
zgc:101021
Chromosome Location
Chr.11: 45126408 - 45138857 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003595.2 NP_001003595.1 2 - 3 822 57
XM_009306324.3 XP_009304599.1 2 - 3 782 57
XM_009306325.3 XP_009304600.1 2 - 3 769 57
GenBank mRNA BC078222.1 - 2 - 3 823 57

View Full Product Details