Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
apip
Entrez Gene ID
447941
Gene Name
APAF1 interacting protein
Gene Aliases
zgc:103619
Chromosome Location
Chr.7: 49901466 - 49908518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004679.1 NP_001004679.1 5 - 6 575 132
XM_005166468.4 XP_005166525.1 5 - 6 586 132
GenBank mRNA BC081498.1 - 5 - 6 575 132
BC165092.1 - 5 - 6 490 132

View Full Product Details