Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
gpsm2l
Entrez Gene ID
449665
Gene Name
G protein signaling modulator 2, like
Gene Aliases
Pinsb, zgc:92571
Chromosome Location
Chr.23: 40133136 - 40189675 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005936.1 NP_001005936.1 5 - 6 774 74
XM_021469788.1 XP_021325463.1 5 - 6 74
GenBank mRNA BC083520.1 - 5 - 6 774 74
DQ235092.1 - 4 - 5 621 74

View Full Product Details