Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sinhcaf
Entrez Gene ID
563991
Gene Name
SIN3-HDAC complex associated factor
Gene Aliases
fam60a, si:ch211-11c20.3
Chromosome Location
Chr.4: 16635446 - 16644708 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025531.1 NP_001020702.1 4 - 5 381 66
GenBank mRNA BC162866.1 - 4 - 5 381 66
BC162884.1 - 4 - 5 381 66

View Full Product Details