The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030172.1 | NP_001025343.1 | 2 - 3 | 454 | 82 |
GenBank mRNA | BC097141.1 | - | 2 - 3 | 454 | 82 |