Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fen1
Entrez Gene ID
386707
Gene Name
flap structure-specific endonuclease 1
Gene Aliases
FEN-1, cb879
Chromosome Location
Chr.24: 11908466 - 11919294 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328507.1 NP_001315436.1 9 - 10 923 110
XM_017353804.1 XP_017209293.1 9 - 10 941 110
GenBank mRNA AY391423.1 - 9 - 10 852 110
BC049413.1 - 9 - 10 1140 110
BC071488.1 - 9 - 10 884 110
CT678576.2 - 3 - 4 520 110

View Full Product Details