Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fastkd1
Entrez Gene ID
100148546
Gene Name
FAST kinase domains 1
Gene Aliases
zgc:77397
Chromosome Location
Chr.9: 48232528 - 48266161 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205629.2 NP_991192.2 6 - 7 1341 74
XM_021478594.1 XP_021334269.1 6 - 7 1358 74
GenBank mRNA BC065356.1 - 6 - 7 1314 74
BC154210.1 - 6 - 7 1220 74

View Full Product Details