Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
st14b
Entrez Gene ID
777629
Gene Name
ST14 transmembrane serine protease matriptase b
Gene Aliases
zgc:152947
Chromosome Location
Chr.21: 25275688 - 25295131 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077737.1 NP_001071205.1 5 - 6 878 84
XM_005161204.4 XP_005161261.1 6 - 7 834 84
XM_009295332.3 XP_009293607.1 6 - 7 754 84
GenBank mRNA BC125837.1 - 5 - 6 878 84
BC165661.1 - 5 - 6 665 84

View Full Product Details