Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
impdh1b
Entrez Gene ID
338306
Gene Name
IMP (inosine 5'-monophosphate) dehydrogenase 1b
Gene Aliases
D3, IMPD 1b, IMPDH 1b, IMPDH1, id:ibd5035, si:dkey-31f5.7, wu:fa09h11, wu:fa99c03, zgc:113446
Chromosome Location
Chr.4: 18960587 - 18985481 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014369.2 NP_001014391.1 9 - 10 1162 77
XM_005164717.4 XP_005164774.1 10 - 11 1668 77
XM_005164718.4 XP_005164775.1 9 - 10 1590 77
XM_017354952.2 XP_017210441.1 10 - 11 1258 77
XM_017354953.2 XP_017210442.1 4 - 5 524 77
GenBank mRNA BC091790.1 - 9 - 10 1084 77
DN896992.1 - 1 - 2 107 77

View Full Product Details