Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nphs1
Entrez Gene ID
692352
Gene Name
NPHS1 adhesion molecule, nephrin
Gene Aliases
nphs1l
Chromosome Location
Chr.15: 36556158 - 36739438 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040687.1 NP_001035777.1 12 - 13 1585 83
XM_017351014.2 XP_017206503.1 14 - 15 2177 83
XM_021481465.1 XP_021337140.1 13 - 14 2061 83
GenBank mRNA DQ466904.1 - 12 - 13 1585 83

View Full Product Details