Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekhg7
Entrez Gene ID
562798
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 7
Gene Aliases
si:ch211-276p10.1, si:dkey-196d8.2, zgc:161981
Chromosome Location
Chr.18: 15887821 - 15932699 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328211.1 NP_001315140.1 14 - 15 2273 148
XM_009293507.3 XP_009291782.1 14 - 15 2467 148
GenBank mRNA BC124825.1 - 8 - 9 879 148

View Full Product Details