Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
foxo1a
Entrez Gene ID
768121
Gene Name
forkhead box O1 a
Gene Aliases
FoxO1a.1, foxo1, zgc:153388
Chromosome Location
Chr.15: 3284004 - 3379044 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077257.2 NP_001070725.2 2 - 3 751 83
XM_009291427.3 XP_009289702.1 2 - 3 1431 83
GenBank mRNA EF421424.1 - 2 - 3 751 83

View Full Product Details