Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sfrp5
Entrez Gene ID
117510
Gene Name
secreted frizzled-related protein 5
Gene Aliases
D144, frp1, zgc:101849
Chromosome Location
Chr.13: 39911551 - 39947335 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131858.1 NP_571933.1 1 - 2 720 101
GenBank mRNA AB055672.1 - 1 - 2 240 101
AY050560.1 - 1 - 2 597 101
BC085455.1 - 1 - 2 720 101
BC164265.1 - 1 - 2 554 101

View Full Product Details