Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stmn1b
Entrez Gene ID
550548
Gene Name
stathmin 1b
Gene Aliases
fj38b07, fj43c10, wu:fj38b07, wu:fj43c10, zgc:110159
Chromosome Location
Chr.16: 34523538 - 34527206 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017850.1 NP_001017850.1 5 - 5 950 152
GenBank mRNA BC092763.1 - 5 - 5 950 152

View Full Product Details