Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tctex1d1
Entrez Gene ID
447884
Gene Name
Tctex1 domain containing 1
Gene Aliases
zgc:101774
Chromosome Location
Chr.2: 14208158 - 14272080 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004623.2 NP_001004623.1 5 - 5 652 146
GenBank mRNA BC081403.1 - 5 - 5 653 146

View Full Product Details