Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mphosph6
Entrez Gene ID
445224
Gene Name
M-phase phosphoprotein 6
Gene Aliases
zgc:100960
Chromosome Location
Chr.25: 35952047 - 35956342 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003618.1 NP_001003618.1 2 - 3 256 73
GenBank mRNA BC078314.1 - 2 - 3 256 73

View Full Product Details