Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2k16
Entrez Gene ID
449790
Gene Name
cytochrome P450, family 2, subfamily K, polypeptide16
Gene Aliases
si:ch211-155i15.2, zgc:101876
Chromosome Location
Chr.3: 12764894 - 12780758 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005963.2 NP_001005963.2 3 - 4 576 68
GenBank mRNA BC083304.1 - 3 - 4 519 68
BC165360.1 - 3 - 4 541 68

View Full Product Details