Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2k16
Entrez Gene ID
449790
Gene Name
cytochrome P450, family 2, subfamily K, polypeptide16
Gene Aliases
si:ch211-155i15.2, zgc:101876
Chromosome Location
Chr.3: 12764894 - 12780758 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005963.2 NP_001005963.2 7 - 8 1247 87
GenBank mRNA BC083304.1 - 7 - 8 1190 87
BC165360.1 - 7 - 8 1212 87

View Full Product Details