Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nr3c2
Entrez Gene ID
562171
Gene Name
nuclear receptor subfamily 3, group C, member 2
Gene Aliases
mr, si:ch211-189l17.1
Chromosome Location
Chr.1: 23728 - 181072 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100403.1 NP_001093873.1 5 - 6 2657 61
GenBank mRNA DQ017617.1 - 3 - 4 470 61
EF567113.1 - 5 - 6 2657 61

View Full Product Details