Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rps23
Entrez Gene ID
100000341
Gene Name
ribosomal protein S23
Gene Aliases
zgc:171710
Chromosome Location
Chr.11: 42556465 - 42561211 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110121.1 NP_001103591.1 2 - 3 198 80
GenBank mRNA BC153998.1 - 2 - 3 198 80
BC171649.1 - 2 - 3 195 80
BC171653.1 - 2 - 3 195 80

View Full Product Details