Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mylpfa
Entrez Gene ID
30429
Gene Name
myosin light chain, phosphorylatable, fast skeletal muscle a
Gene Aliases
cb46, mlc2f, mylz2, sb:cb46
Chromosome Location
Chr.3: 294135 - 299623 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131188.2 NP_571263.1 5 - 6 420 85
GenBank mRNA AF081462.1 - 5 - 6 406 85
BC045520.1 - 5 - 6 420 85
BC163956.1 - 4 - 5 399 85

View Full Product Details