Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pex3
Entrez Gene ID
393197
Gene Name
peroxisomal biogenesis factor 3
Gene Aliases
zgc:56313
Chromosome Location
Chr.20: 37760493 - 37769303 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200228.1 NP_956522.1 1 - 2 217 103
GenBank mRNA BC046066.1 - 1 - 2 217 103

View Full Product Details