Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pam16
Entrez Gene ID
393777
Gene Name
presequence translocase associated motor 16
Gene Aliases
magmas, timm16, zgc:73356
Chromosome Location
Chr.3: 12345276 - 12352795 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200804.2 NP_957098.2 2 - 3 235 79
GenBank mRNA BC059670.1 - 2 - 3 183 79

View Full Product Details