Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ca7
Entrez Gene ID
564201
Gene Name
carbonic anhydrase VII
Gene Aliases
zgc:73385
Chromosome Location
Chr.7: 68551 - 79855 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200813.1 NP_957107.1 3 - 4 448 91
GenBank mRNA BC059693.1 - 3 - 4 448 91
BC154317.1 - 3 - 4 552 91
BC171412.1 - 3 - 4 407 91
BC171416.1 - 3 - 4 407 91

View Full Product Details