Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nudcd3
Entrez Gene ID
406472
Gene Name
NudC domain containing 3
Gene Aliases
zgc:77067
Chromosome Location
Chr.10: 45059797 - 45070182 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213191.1 NP_998356.1 1 - 2 291 56
GenBank mRNA BC065992.1 - 1 - 2 291 56

View Full Product Details