Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGFR
Entrez Gene ID
100009714
Gene Name
prostaglandin F receptor
Gene Aliases
-
Chromosome Location
Chr.5: 81328928 - 81377382 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081806.3 NP_001075275.1 1 - 2 822 85
XM_005610256.3 XP_005610313.1 - 968 85
XM_014739993.2 XP_014595479.1 - 1122 85
XM_023640207.1 XP_023495975.1 - 1212 85
XM_023640208.1 XP_023495976.1 - 1000 85
GenBank mRNA DQ385610.1 - 1 - 2 822 85

View Full Product Details