Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCER2
Entrez Gene ID
100009715
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
CD23
Chromosome Location
Chr.7: 5474789 - 5484667 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081807.2 NP_001075276.1 10 - 11 771 106
XM_023644108.1 XP_023499876.1 - 815 106
XM_023644110.1 XP_023499878.1 - 761 106
XM_023644111.1 XP_023499879.1 - 707 106
XM_023644112.1 XP_023499880.1 - 704 106
GenBank mRNA AF141931.1 - 10 - 11 779 106

View Full Product Details