Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P19
Entrez Gene ID
100034213
Gene Name
P19 lipocalin
Gene Aliases
-
Chromosome Location
Chr.25: 38802060 - 38805847 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082509.2 NP_001075978.2 3 - 4 315 84
GenBank mRNA X98459.1 - - 321 84

View Full Product Details