Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD1
Entrez Gene ID
100033850
Gene Name
SMAD family member 1
Gene Aliases
-
Chromosome Location
Chr.2: 87271832 - 87351742 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163882.1 NP_001157354.1 6 - 7 1503 93
XM_005607767.3 XP_005607824.1 - 1698 93
XM_005607768.3 XP_005607825.1 - 3647 93
XM_005607769.3 XP_005607826.1 - 1769 93
XM_005607770.3 XP_005607827.1 - 1574 93
XM_023626119.1 XP_023481887.1 - 1736 93
XM_023626135.1 XP_023481903.1 - 1897 93
XM_023626149.1 XP_023481917.1 - 1363 93
GenBank mRNA AB106113.1 - 1 - 2 92 93

View Full Product Details