Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM6
Entrez Gene ID
100568303
Gene Name
tripartite motif containing 6
Gene Aliases
-
Chromosome Location
Chr.7: 76586899 - 76604032 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205192.1 NP_001192121.1 5 - 6 915 83
XM_023644173.1 XP_023499941.1 - 1456 83

View Full Product Details