Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR30D
- Entrez Gene ID
-
100314834
- Gene Name
- microRNA mir-30d
- Gene Aliases
-
eca-mir-30d
- Chromosome Location
-
Chr.9: 77319227 - 77319297 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_032886.1 |
|
1 - 1 |
30 |
66 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA mir-30b |
MIR30B |
eca-mir-30b |
-4.3kb
|
Sense |
Chr. - 77314900 - 77314957 on Build EquCab3.0 |
Downstream |
KH RNA binding domain containing, signal transduction associated 3 |
KHDRBS3 |
|
502.8kb
|
Anti-sense |
Chr. - 77822133 - 78012097 on Build EquCab3.0 |
View Full Product Details